Assay Details
| Gene Symbol: Ac78C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259928.2 | NP_001246857.1 | 4 - 5 | 590 | 78 | |
| NM_001275208.1 | NP_001262137.1 | 10 - 11 | 2389 | 78 | ||
| NM_001275209.1 | NP_001262138.1 | 11 - 12 | 2600 | 78 | ||
| NM_001275210.1 | NP_001262139.1 | 11 - 12 | 2597 | 78 | ||
| NM_079470.5 | NP_524194.3 | 11 - 12 | 2600 | 78 | ||
More Information
Set Membership: |
|

Ac78C
Genomic Map