Assay Details
| Gene Symbol: Ac78C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259928.2 | NP_001246857.1 | 5 - 6 | 727 | 95 | |
| NM_001275208.1 | NP_001262137.1 | 11 - 12 | 2526 | 95 | ||
| NM_001275209.1 | NP_001262138.1 | 12 - 13 | 2737 | 95 | ||
| NM_001275210.1 | NP_001262139.1 | 12 - 13 | 2734 | 95 | ||
| NM_079470.5 | NP_524194.3 | 12 - 13 | 2737 | 95 | ||
More Information
Set Membership: |
|

Ac78C
Genomic Map