Assay Details
| Gene Symbol: Ac78C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275208.1 | NP_001262137.1 | 2 - 3 | 877 | 76 | |
| NM_001275209.1 | NP_001262138.1 | 3 - 4 | 1088 | 76 | ||
| NM_079470.5 | NP_524194.3 | 3 - 4 | 1088 | 76 | ||
More Information
Set Membership: |
|

Ac78C
Genomic Map