Assay Details
| Gene Symbol: Ac78C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259928.2 | NP_001246857.1 | 1 - 2 | 206 | 68 | |
| NM_001275208.1 | NP_001262137.1 | 7 - 8 | 2005 | 68 | ||
| NM_001275209.1 | NP_001262138.1 | 8 - 9 | 2216 | 68 | ||
| NM_001275210.1 | NP_001262139.1 | 8 - 9 | 2213 | 68 | ||
| NM_079470.5 | NP_524194.3 | 8 - 9 | 2216 | 68 | ||
More Information
Set Membership: |
|

Ac78C
Genomic Map