Assay Details
| Gene Symbol: Pc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079475.3 | NP_524199.1 | 1 - 2 | 228 | 66 | |
| GenBank mRNA | AY071619.2 | - | 1 - 2 | 183 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pc| Gene Symbol: Pc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079475.3 | NP_524199.1 | 1 - 2 | 228 | 66 | |
| GenBank mRNA | AY071619.2 | - | 1 - 2 | 183 | 66 | |
Set Membership: |
|