Assay Details
| Gene Symbol: CycH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079483.3 | NP_524207.1 | 1 - 2 | 326 | 92 | |
| GenBank mRNA | AF074334.1 | - | 2 - 3 | 397 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CycH| Gene Symbol: CycH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079483.3 | NP_524207.1 | 1 - 2 | 326 | 92 | |
| GenBank mRNA | AF074334.1 | - | 2 - 3 | 397 | 92 | |
Set Membership: |
|