Assay Details
| Gene Symbol: alpha-Cat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300225.1 | NP_001287154.1 | 7 - 8 | 2589 | 117 | |
| NM_079495.3 | NP_524219.1 | 8 - 9 | 2102 | 117 | ||
| GenBank mRNA | D13964.2 | - | 8 - 9 | 2065 | 117 | |
More Information
Set Membership: |
|

alpha-Cat
Genomic Map