Assay Details
| Gene Symbol: Ser8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137030.3 | NP_610874.1 | 1 | 743 | 57 | |
| GenBank mRNA | BT023280.1 | - | 1 | 743 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ser8| Gene Symbol: Ser8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137030.3 | NP_610874.1 | 1 | 743 | 57 | |
| GenBank mRNA | BT023280.1 | - | 1 | 743 | 57 | |
Set Membership: |
|