Assay Details
| Gene Symbol: mars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169671.2 | NP_001163142.1 | 2 - 3 | 2412 | 71 | |
| NM_001299458.1 | NP_001286387.1 | 2 - 3 | 2127 | 71 | ||
| NM_001299459.1 | NP_001286388.1 | 3 - 4 | 3052 | 71 | ||
| NM_137034.4 | NP_610878.1 | 1 - 2 | 1613 | 71 | ||
| GenBank mRNA | AJ565928.1 | - | 1 - 2 | 1600 | 71 | |
| AY058511.1 | - | 1 - 2 | 1601 | 71 | ||
More Information
Set Membership: |
|

mars
Genomic Map