Assay Details
| Gene Symbol: S-Lap7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274030.1 | NP_001260959.1 | 1 | 1949 | 116 | |
| NM_137056.2 | NP_610900.1 | 1 | 1949 | 116 | ||
| GenBank mRNA | BT022093.1 | - | 1 | 1948 | 116 | |
More Information
Set Membership: |
|

S-Lap7
Genomic Map