Assay Details
| Gene Symbol: Ctf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137077.4 | NP_610921.1 | 1 | 2753 | 130 | |
| GenBank mRNA | BT023774.1 | - | 1 | 2707 | 130 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctf4| Gene Symbol: Ctf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137077.4 | NP_610921.1 | 1 | 2753 | 130 | |
| GenBank mRNA | BT023774.1 | - | 1 | 2707 | 130 | |
Set Membership: |
|