Assay Details
| Gene Symbol: Usp20-33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137099.3 | NP_610943.2 | 2 - 3 | 551 | 70 | |
| GenBank mRNA | AY118338.1 | - | 2 - 3 | 542 | 70 | |
More Information
Set Membership: |
|

Usp20-33
Genomic Map