Assay Details
| Gene Symbol: tej | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137106.3 | NP_610950.2 | 1 - 2 | 298 | 75 | |
| GenBank mRNA | BT030811.1 | - | 1 - 2 | 298 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tej| Gene Symbol: tej | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137106.3 | NP_610950.2 | 1 - 2 | 298 | 75 | |
| GenBank mRNA | BT030811.1 | - | 1 - 2 | 298 | 75 | |
Set Membership: |
|