Assay Details
| Gene Symbol: tej | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137106.3 | NP_610950.2 | 5 - 6 | 1674 | 130 | |
| GenBank mRNA | BT030811.1 | - | 5 - 6 | 1674 | 130 | |
More Information
Set Membership: |
|

tej
Genomic Map