Assay Details
| Gene Symbol: Arc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137112.2 | NP_610956.1 | 1 | 653 | 154 | |
| GenBank mRNA | AY119191.1 | - | 1 | 633 | 154 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arc2| Gene Symbol: Arc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137112.2 | NP_610956.1 | 1 | 653 | 154 | |
| GenBank mRNA | AY119191.1 | - | 1 | 633 | 154 | |
Set Membership: |
|