Assay Details
| Gene Symbol: Adgf-E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137133.2 | NP_610977.1 | 1 - 2 | 1096 | 90 | |
| GenBank mRNA | AF337551.1 | - | 1 - 2 | 1092 | 90 | |
| BT150197.1 | - | 1 - 2 | 1096 | 90 | ||
More Information
Set Membership: |
|

Adgf-E
Genomic Map