Assay Details
| Gene Symbol: Lap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202009.2 | NP_001188938.1 | 3 - 4 | 706 | 84 | |
| NM_137163.5 | NP_611007.1 | 3 - 4 | 689 | 84 | ||
| GenBank mRNA | BT011365.1 | - | 3 - 4 | 690 | 84 | |
More Information
Set Membership: |
|

Lap1
Genomic Map