Assay Details
| Gene Symbol: GPHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137172.4 | NP_611016.2 | 3 - 4 | 706 | 84 | |
| GenBank mRNA | BT023838.1 | - | 3 - 4 | 682 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPHR| Gene Symbol: GPHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137172.4 | NP_611016.2 | 3 - 4 | 706 | 84 | |
| GenBank mRNA | BT023838.1 | - | 3 - 4 | 682 | 84 | |
Set Membership: |
|