Assay Details
| Gene Symbol: ItgaPS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137181.3 | NP_611025.2 | 5 - 6 | 1941 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ItgaPS4| Gene Symbol: ItgaPS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137181.3 | NP_611025.2 | 5 - 6 | 1941 | 59 | |
Set Membership: |
|