Assay Details
Gene Symbol: unc-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299521.1 | NP_001286450.1 | 4 - 5 | 1842 | 63 | |
NM_137189.3 | NP_611033.2 | 3 - 4 | 1640 | 63 | ||
GenBank mRNA | AY058501.1 | - | 3 - 4 | 1614 | 63 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
