Assay Details
| Gene Symbol: Zasp52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032249.4 | NP_001027420.2 | 10 - 11 | 1704 | 67 | |
| NM_001043086.3 | NP_001036551.1 | 10 - 11 | 1878 | 67 | ||
| NM_001274076.1 | NP_001261005.1 | 10 - 11 | 1704 | 67 | ||
| NM_001274081.1 | NP_001261010.1 | 2 - 3 | 613 | 67 | ||
| NM_001274086.1 | NP_001261015.1 | 10 - 11 | 1878 | 67 | ||
| NM_001316422.1 | NP_001303351.1 | 2 - 3 | 613 | 67 | ||
| NM_137214.5 | NP_611058.4 | 10 - 11 | 2454 | 67 | ||
| NM_145757.3 | NP_665700.2 | 10 - 11 | 2510 | 67 | ||
| GenBank mRNA | BT022171.2 | - | 10 - 11 | 1858 | 67 | |
| JN034429.1 | - | 10 - 11 | 1865 | 67 | ||
| JN034430.1 | - | 2 - 3 | 533 | 67 | ||
More Information
Set Membership: |
|

Zasp52
Genomic Map