Assay Details
| Gene Symbol: CG8399 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299543.1 | NP_001286472.1 | 4 - 5 | 1101 | 86 | |
| NM_137235.3 | NP_611079.2 | 4 - 5 | 1101 | 86 | ||
| GenBank mRNA | AY118595.1 | - | 4 - 5 | 1083 | 86 | |
More Information
Set Membership: |
|

CG8399
Genomic Map