Assay Details
| Gene Symbol: CG8399 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137235.3 | NP_611079.2 | 6 - 7 | 1835 | 68 | |
| GenBank mRNA | AY118595.1 | - | 6 - 7 | 1817 | 68 | |
More Information
Set Membership: |
|

CG8399
Genomic Map