Assay Details
| Gene Symbol: CG43689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272298.2 | NP_001259227.1 | 3 - 4 | 3258 | 63 | |
| NM_001272299.2 | NP_001259228.1 | 5 - 6 | 3501 | 63 | ||
| NM_001272300.2 | NP_001259229.1 | 3 - 4 | 3258 | 63 | ||
| NM_001272301.2 | NP_001259230.1 | 4 - 5 | 3432 | 63 | ||
| NM_001297921.1 | NP_001284850.1 | 4 - 5 | 3168 | 63 | ||
| NM_001370021.1 | NP_001356932.1 | 3 - 4 | 3493 | 63 | ||
| NM_167000.4 | NP_726905.2 | 4 - 5 | 3432 | 63 | ||
More Information
Set Membership: |
|

CG43689
Genomic Map