Assay Details
| Gene Symbol: CG43689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272298.2 | NP_001259227.1 | 4 - 5 | 3527 | 76 | |
| NM_001272299.2 | NP_001259228.1 | 6 - 7 | 3770 | 76 | ||
| NM_001272300.2 | NP_001259229.1 | 4 - 5 | 3527 | 76 | ||
| NM_001272301.2 | NP_001259230.1 | 5 - 6 | 3701 | 76 | ||
| NM_001297921.1 | NP_001284850.1 | 5 - 6 | 3437 | 76 | ||
| NM_001370021.1 | NP_001356932.1 | 4 - 5 | 3762 | 76 | ||
| NM_167000.4 | NP_726905.2 | 5 - 6 | 3701 | 76 | ||
More Information
Set Membership: |
|

CG43689
Genomic Map