Assay Details
| Gene Symbol: norpA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014720.2 | NP_001014720.1 | 13 - 14 | 4110 | 70 | |
| NM_001014721.3 | NP_001014721.1 | 12 - 13 | 3648 | 70 | ||
| NM_001169190.2 | NP_001162661.1 | 14 - 15 | 4379 | 70 | ||
| NM_001297931.1 | NP_001284860.1 | 13 - 14 | 3773 | 70 | ||
| NM_080330.4 | NP_525069.2 | 12 - 13 | 3648 | 70 | ||
| NM_167008.2 | NP_726925.1 | 13 - 14 | 4110 | 70 | ||
| GenBank mRNA | AF181641.1 | - | 12 - 13 | 3631 | 70 | |
More Information
Set Membership: |
|

norpA
Genomic Map