Assay Details
| Gene Symbol: norpA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169190.2 | NP_001162661.1 | 6 - 7 | 1637 | 66 | |
| NM_080330.4 | NP_525069.2 | 4 - 5 | 906 | 66 | ||
| NM_167008.2 | NP_726925.1 | 5 - 6 | 1368 | 66 | ||
| GenBank mRNA | AF181641.1 | - | 4 - 5 | 889 | 66 | |
More Information
Set Membership: |
|

norpA
Genomic Map