Assay Details
| Gene Symbol: fzr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080113.3 | NP_524852.2 | 4 - 5 | 1076 | 71 | |
| NM_167013.2 | NP_726941.1 | 4 - 5 | 982 | 71 | ||
| GenBank mRNA | BT001403.1 | - | 4 - 5 | 1668 | 71 | |
| BT010011.1 | - | 4 - 5 | 1126 | 71 | ||
More Information
Set Membership: |
|

fzr
Genomic Map