Assay Details
| Gene Symbol: MAPk-Ak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201618.1 | NP_001188547.1 | 3 - 4 | 1147 | 63 | |
| NM_001258607.2 | NP_001245536.1 | 2 - 3 | 1000 | 63 | ||
| NM_080030.5 | NP_524769.1 | 3 - 4 | 1107 | 63 | ||
| NM_176688.3 | NP_788861.1 | 3 - 4 | 1132 | 63 | ||
| GenBank mRNA | BT125824.1 | - | 3 - 4 | 1045 | 63 | |
| U20757.1 | - | 3 - 4 | 1074 | 63 | ||
More Information
Set Membership: |
|

MAPk-Ak2
Genomic Map