Assay Details
| Gene Symbol: kdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132091.3 | NP_572319.2 | 2 - 3 | 1180 | 96 | |
| NM_167072.2 | NP_727091.1 | 2 - 3 | 1306 | 96 | ||
| GenBank mRNA | AY089318.1 | - | 2 - 3 | 1306 | 96 | |
More Information
Set Membership: |
|

kdn
Genomic Map