Assay Details
| Gene Symbol: APC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132101.4 | NP_572329.3 | 2 - 3 | 617 | 78 | |
| NM_167079.2 | NP_727101.1 | 1 - 2 | 592 | 78 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

APC7| Gene Symbol: APC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132101.4 | NP_572329.3 | 2 - 3 | 617 | 78 | |
| NM_167079.2 | NP_727101.1 | 1 - 2 | 592 | 78 | ||
Set Membership: |
|