Assay Details
| Gene Symbol: APC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132101.4 | NP_572329.3 | 4 - 5 | 1567 | 83 | |
| NM_167079.2 | NP_727101.1 | 3 - 4 | 1542 | 83 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

APC7| Gene Symbol: APC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132101.4 | NP_572329.3 | 4 - 5 | 1567 | 83 | |
| NM_167079.2 | NP_727101.1 | 3 - 4 | 1542 | 83 | ||
Set Membership: |
|