Assay Details
| Gene Symbol: Cdc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298021.1 | NP_001284950.1 | 3 - 4 | 861 | 56 | |
| NM_167080.2 | NP_727103.1 | 3 - 4 | 1002 | 56 | ||
More Information
Set Membership: |
|

Cdc7
Genomic Map