Assay Details
| Gene Symbol: sov | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298033.1 | NP_001284962.1 | 5 - 6 | 8317 | 91 | |
| NM_132120.2 | NP_572348.1 | 5 - 6 | 8541 | 91 | ||
| NM_167090.2 | NP_727123.1 | 5 - 6 | 8320 | 91 | ||
More Information
Set Membership: |
|

sov
Genomic Map