Assay Details
| Gene Symbol: sov | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298033.1 | NP_001284962.1 | 6 - 7 | 8960 | 56 | |
| NM_132120.2 | NP_572348.1 | 6 - 7 | 9184 | 56 | ||
| NM_167090.2 | NP_727123.1 | 6 - 7 | 8963 | 56 | ||
More Information
Set Membership: |
|

sov
Genomic Map