Assay Details
| Gene Symbol: nonC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298038.1 | NP_001284967.1 | 8 - 9 | 9605 | 67 | |
| NM_167095.3 | NP_727132.1 | 8 - 9 | 9605 | 67 | ||
More Information
Set Membership: |
|

nonC
Genomic Map