Assay Details
| Gene Symbol: Moe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258654.1 | NP_001245583.1 | 4 - 5 | 602 | 77 | |
| NM_001272432.1 | NP_001259361.1 | 5 - 6 | 533 | 77 | ||
| NM_167168.4 | NP_727290.1 | 4 - 5 | 602 | 77 | ||
| NM_167169.2 | NP_727291.1 | 4 - 5 | 493 | 77 | ||
| GenBank mRNA | BT099729.1 | - | 4 - 5 | 602 | 77 | |
More Information
Set Membership: |
|

Moe
Genomic Map