Assay Details
| Gene Symbol: Obp8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_167184.2 | NP_727322.1 | 1 - 2 | 63 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Obp8a| Gene Symbol: Obp8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_167184.2 | NP_727322.1 | 1 - 2 | 63 | 86 | |
Set Membership: |
|