Assay Details
| Gene Symbol: Hex-A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272455.2 | NP_001259384.1 | 1 | 2359 | 93 | |
| NM_080109.4 | NP_524848.1 | 1 | 2144 | 93 | ||
| NM_167194.3 | NP_727350.1 | 1 | 2144 | 93 | ||
| GenBank mRNA | AY069391.1 | - | 1 | 2149 | 93 | |
More Information
Set Membership: |
|

Hex-A
Genomic Map