Assay Details
| Gene Symbol: ZAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258677.2 | NP_001245606.1 | 5 - 6 | 4227 | 68 | |
| NM_001258678.2 | NP_001245607.1 | 10 - 11 | 5689 | 68 | ||
| NM_167218.2 | NP_727393.1 | 10 - 11 | 5686 | 68 | ||
More Information
Set Membership: |
|

ZAP3
Genomic Map