Assay Details
| Gene Symbol: ZAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258678.2 | NP_001245607.1 | 6 - 7 | 5216 | 73 | |
| NM_167218.2 | NP_727393.1 | 6 - 7 | 5213 | 73 | ||
More Information
Set Membership: |
|

ZAP3
Genomic Map