Assay Details
| Gene Symbol: ZAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258677.2 | NP_001245606.1 | 3 - 4 | 4058 | 75 | |
| NM_001258678.2 | NP_001245607.1 | 8 - 9 | 5520 | 75 | ||
| NM_167218.2 | NP_727393.1 | 8 - 9 | 5517 | 75 | ||
More Information
Set Membership: |
|

ZAP3
Genomic Map