Assay Details
| Gene Symbol: CG10353 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258704.1 | NP_001245633.1 | 5 - 6 | 2201 | 69 | |
| NM_001272540.1 | NP_001259469.1 | 7 - 8 | 2124 | 69 | ||
| NM_001272541.2 | NP_001259470.2 | 5 - 6 | 1922 | 69 | ||
| NM_001272543.1 | NP_001259472.1 | 7 - 8 | 2403 | 69 | ||
| NM_001272544.2 | NP_001259473.2 | 5 - 6 | 2114 | 69 | ||
| NM_132534.4 | NP_572762.2 | 5 - 6 | 1835 | 69 | ||
| NM_167312.4 | NP_727578.3 | 6 - 7 | 2406 | 69 | ||
| GenBank mRNA | BT099944.1 | - | 5 - 6 | 1832 | 69 | |
More Information
Set Membership: |
|

CG10353
Genomic Map