Assay Details
| Gene Symbol: Tomosyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169264.2 | NP_001162735.1 | 18 - 19 | 4383 | 66 | |
| NM_001169265.2 | NP_001162736.2 | 18 - 19 | 4134 | 66 | ||
| NM_001258715.2 | NP_001245644.1 | 18 - 19 | 3735 | 66 | ||
| NM_132591.5 | NP_572819.4 | 19 - 20 | 3780 | 66 | ||
| NM_167328.4 | NP_727628.3 | 19 - 20 | 4423 | 66 | ||
| NM_167329.3 | NP_727629.2 | 18 - 19 | 4583 | 66 | ||
| NM_167330.3 | NP_727630.3 | 18 - 19 | 4256 | 66 | ||
More Information
Set Membership: |
|

Tomosyn
Genomic Map