Assay Details
| Gene Symbol: Tomosyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169264.2 | NP_001162735.1 | 9 - 10 | 1688 | 71 | |
| NM_001169265.2 | NP_001162736.2 | 9 - 10 | 1688 | 71 | ||
| NM_001258715.2 | NP_001245644.1 | 9 - 10 | 1688 | 71 | ||
| NM_132591.5 | NP_572819.4 | 10 - 11 | 1733 | 71 | ||
| NM_167328.4 | NP_727628.3 | 10 - 11 | 1728 | 71 | ||
| NM_167329.3 | NP_727629.2 | 9 - 10 | 1888 | 71 | ||
| NM_167330.3 | NP_727630.3 | 9 - 10 | 1810 | 71 | ||
More Information
Set Membership: |
|

Tomosyn
Genomic Map