Assay Details
| Gene Symbol: Tomosyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169264.2 | NP_001162735.1 | 14 - 15 | 3568 | 92 | |
| NM_167328.4 | NP_727628.3 | 15 - 16 | 3608 | 92 | ||
| NM_167329.3 | NP_727629.2 | 14 - 15 | 3768 | 92 | ||
More Information
Set Membership: |
|

Tomosyn
Genomic Map