Assay Details
| Gene Symbol: sv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014697.2 | NP_001014697.1 | 11 - 12 | 2358 | 73 | |
| NM_001014699.3 | NP_001014699.1 | 11 - 12 | 2550 | 73 | ||
| NM_079894.4 | NP_524633.3 | 11 - 12 | 2550 | 73 | ||
| GenBank mRNA | AF010256.1 | - | 11 - 12 | 2530 | 73 | |
More Information
Set Membership: |
|

sv
Genomic Map