Assay Details
| Gene Symbol: toy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272151.1 | NP_001259080.1 | 7 - 8 | 1811 | 81 | |
| NM_001382011.1 | NP_001368990.1 | 7 - 8 | 1862 | 81 | ||
| NM_079899.4 | NP_524638.3 | 7 - 8 | 1862 | 81 | ||
| GenBank mRNA | AY051487.1 | - | 7 - 8 | 1834 | 81 | |
More Information
Set Membership: |
|

toy
Genomic Map