Assay Details
| Gene Symbol: nej | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201646.2 | NP_001188575.1 | 12 - 13 | 9134 | 68 | |
| NM_001201647.2 | NP_001188576.1 | 12 - 13 | 9086 | 68 | ||
| NM_001272458.1 | NP_001259387.1 | 12 - 13 | 8988 | 68 | ||
| NM_079903.4 | NP_524642.2 | 12 - 13 | 9163 | 68 | ||
More Information
Set Membership: |
|

nej
Genomic Map