Assay Details
| Gene Symbol: syd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169902.2 | NP_001163373.1 | 7 - 8 | 1701 | 64 | |
| NM_001274622.1 | NP_001261551.1 | 6 - 7 | 1680 | 64 | ||
| NM_001274623.1 | NP_001261552.1 | 6 - 7 | 1680 | 64 | ||
| NM_001274624.1 | NP_001261553.1 | 6 - 7 | 1680 | 64 | ||
| NM_079913.3 | NP_524652.1 | 7 - 8 | 1701 | 64 | ||
| NM_168245.3 | NP_729311.2 | 7 - 8 | 1126 | 64 | ||
| GenBank mRNA | AF262045.1 | - | 7 - 8 | 1455 | 64 | |
More Information
Set Membership: |
|

syd
Genomic Map